Learn More
Alkali Scientific Cantharidin, 25 Mg
SDP
Catalog No. 5000661503
Encompass_Preferred
Cantharidin, 25 mg, is a high-purity reagent widely used in research settings, particularly in studies involving cellular biology, histology, and pharmacology. It is a naturally occurring compound that has been extensively studied for its cytotoxic effects on certain cell types, including cancerous cells. Cantharidin's mechanism of action involves inhibiting protein phosphatases, which disrupts cell cycle regulation and induces apoptosis in target cells. This research-grade reagent ensures precise and consistent results, and is often used in studies of cell membrane permeability, cancer cell biology, and therapeutic applications in experimental research, providing valuable insights into cellular mechanisms and drug development.
Catalog No. 50-006-61503 Supplier Alkali Scientific Supplier No. CAN002CP
Cantharidin, 25 mg, is a high-purity reagent widely used in research settings, particularly in studies involving cellular biology, histology, and pharmacology. It is a naturally occurring compound that has been extensively studied for its cytotoxic effects on certain cell types, including cancerous cells. Cantharidin's mechanism of action involves inhibiting protein phosphatases, which disrupts cell cycle regulation and induces apoptosis in target cells. This research-grade reagent ensures precise and consistent results, and is often used in studies of cell membrane permeability, cancer cell biology, and therapeutic applications in experimental research, providing valuable insights into cellular mechanisms and drug development.