Learn More
Enzo Life Sciences Cantharidin (25mg). CAS: 56-25-7
SDP
Supplier:  Enzo Life Sciences ALX270063M025
Encompass_Preferred
Potent protein phosphatase 2A (PP2A) inhibitor. At higher concentrations, also inhibits protein phosphatase 1 (PP1). Alternative name: Hexahydro-3a,7a-dimethyl-4,7-epoxyisobenzo-furan-1,3-dione. Formula: C10H12O4. MW: 196.2. MI: 14: 1752. Purity: ≥98% (GC). Appearance: White to off-white solid. Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: Ambient. Handling: Protect from light.
Catalog No. 50-200-8395