Learn More
Apexbio Technology LLC Canthin-6-one 479-43-6 1mg
SDP
Catalog No. 5000089333
Encompass_Preferred
Canthin-6-one (CAS 479-43-6) is a -carboline alkaloid recognized for its broad-spectrum biological activity In the context of mycobacterial research Canthin-6-one has demonstrated inhibitory effects against Mycobacterium tuberculosis Evidence suggests that its antimicrobial action is likely mediated through disruption of cell wall biosynthesis or interference with metabolic pathways critical for bacterial viability Owing to these properties Canthin-6-one serves as a valuable tool for elucidating antibacterial mechanisms assessing drug susceptibility in resistant strains and exploring candidate molecules for anti-infective drug development
Catalog No. 50-000-89333 Supplier Apexbio Technology LLC Supplier No. BA13741
Canthin-6-one (CAS 479-43-6) is a -carboline alkaloid recognized for its broad-spectrum biological activity In the context of mycobacterial research Canthin-6-one has demonstrated inhibitory effects against Mycobacterium tuberculosis Evidence suggests that its antimicrobial action is likely mediated through disruption of cell wall biosynthesis or interference with metabolic pathways critical for bacterial viability Owing to these properties Canthin-6-one serves as a valuable tool for elucidating antibacterial mechanisms assessing drug susceptibility in resistant strains and exploring candidate molecules for anti-infective drug development