Learn More
Grainger Cantilever Shelf 48 in L 1-1/4 in H
Catalog No. 502768123
Encompass_Preferred
The Pro-Line Cantilever Shelf is a 48 inch wide steel shelf that can hold up to 250 lbs. It has a black powder coat finish and mounting brackets for easy installation. The shelf depth is 12 inches and it's made in the USA. This heavy duty shelf is ideal for workshops, garages, basements, and storage rooms to hold tools, supplies, and equipment.
Catalog No. 50-276-8123 Supplier Grainger Supplier No. 16Y942
The Pro-Line Cantilever Shelf is a 48 inch wide steel shelf that can hold up to 250 lbs. It has a black powder coat finish and mounting brackets for easy installation. The shelf depth is 12 inches and it's made in the USA. This heavy duty shelf is ideal for workshops, garages, basements, and storage rooms to hold tools, supplies, and equipment.