Learn More
Grainger PRO-LINE Cantilever Shelf, Steel, 72 in. x 12 in., Gray, 1 ea
Catalog No. 502790222
Encompass_Preferred
This cantilever shelf provides a durable, flat surface for organized storage and display on compatible workbenches. Designed to maximize workspace efficiency, it keeps essential items within easy reach while helping to clear and declutter work surfaces.
  • Constructed from powder coated steel with 16 ga gauge
  • Features adjustable height for flexible positioning
  • Includes necessary mounting brackets and hardware for installation
  • Compatible with IWB series workbenches and CPB72 units
  • Overall load capacity of 125 lb
  • Overall dimensions of 182.88 cm (72 in.) width by 30.48 cm (12 in.) depth
  • Carbon free and RoHS compliant
Catalog No. 50-279-0222 Supplier Grainger Supplier No. 16D741
This cantilever shelf provides a durable, flat surface for organized storage and display on compatible workbenches. Designed to maximize workspace efficiency, it keeps essential items within easy reach while helping to clear and declutter work surfaces.
  • Constructed from powder coated steel with 16 ga gauge
  • Features adjustable height for flexible positioning
  • Includes necessary mounting brackets and hardware for installation
  • Compatible with IWB series workbenches and CPB72 units
  • Overall load capacity of 125 lb
  • Overall dimensions of 182.88 cm (72 in.) width by 30.48 cm (12 in.) depth
  • Carbon free and RoHS compliant