Learn More
Aviva Systems Biology Corp CANX Antibody - C-terminal region (ARP59755_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59755_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CANX. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CANX. Alternate names: CNX, FLJ26570, IP90, P90ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-813