Learn More
ATCC Caov-4; Ovarian Adenocarcinoma; Human (Homo Sapiens)
Catalog No. 502382970
Encompass
Caov-4 is a cell line exhibiting epithelial morphology that was isolated from the ovaries of a 45-year-old, White female patient with adenocarcinoma. This product has applications in immuno-oncology.
Catalog No. 50-238-2970 Supplier ATCC Supplier No. HTB76
Caov-4 is a cell line exhibiting epithelial morphology that was isolated from the ovaries of a 45-year-old, White female patient with adenocarcinoma. This product has applications in immuno-oncology.