Learn More
Chemglass Life Sciences Cap, 100mm, Blue, Polypropylene, Non-Sterile
SDP
Catalog No. 50143771
Encompass_Preferred
Blue polypropylene cap is fully autoclavable at 121 degreesC.For a tight seal, cap must be used withsilicone liner (listed below).
Catalog No. 50-143-771 Supplier Chemglass Life Sciences Supplier No. CLS1487100
Blue polypropylene cap is fully autoclavable at 121 degreesC.For a tight seal, cap must be used withsilicone liner (listed below).