Learn More
Perkin Elmer US LLC 11 mm Pre-Assembled Red Aluminum Crimp Cap with PTFE/Red Rubber Septa, 100/pk
Catalog No. 509052341
Encompass_Preferred
For sealing sample vials for temperatures up to 160°C. Recommend for storage and for multiple injection use.
Catalog No. 50-905-2341 Supplier Perkin Elmer US LLC Supplier No. N9302685
For sealing sample vials for temperatures up to 160°C. Recommend for storage and for multiple injection use.