Learn More
Foxx Life Sciences Foxx Life Sciences™ VersaCap Filling and Venting closures
Catalog No. NC1984508
Encompass_Preferred
Foxx Life Sciences™ VersaCap Filling and Venting closures are aseptic handling solutions for pharmaceutical, biotechnology, and laboratory liquid transfer applications. All closures can be used on bottles and carboys with 53B, 83B, or 120mm closures. The pre-mounted lengths of platinum-cured silicone tubing allow for easy customization of carboys and bottles.
Catalog No. NC1984508 Supplier Foxx Life Sciences Supplier No. 2056203RLS
Foxx Life Sciences™ VersaCap Filling and Venting closures are aseptic handling solutions for pharmaceutical, biotechnology, and laboratory liquid transfer applications. All closures can be used on bottles and carboys with 53B, 83B, or 120mm closures. The pre-mounted lengths of platinum-cured silicone tubing allow for easy customization of carboys and bottles.