Learn More
Ace Glass, Inc. Caps, 18-400, white polypropylene, PTFE faced silicone liner, open top, SP/100, CS/1000
SDP
Catalog No. 50971989
Encompass
Catalog No. 50-971-989 Supplier Ace Glass, Inc. Supplier No. 570226