Learn More
Perkin Elmer US LLC Pre-Assembled Magnetic Steel Screw Cap with PTFE/Silicone (White) Septa, 100/pk
Catalog No. 509052427
Encompass_Preferred
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.
Catalog No. 50-905-2427 Supplier Perkin Elmer US LLC Supplier No. N9306077
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.