Learn More
Ace Glass, Inc. Caps, 20-400, white polypropylene, PTFE faced silicone liner, closed top, SP/100, CS/1000
SDP
Catalog No. 50971986
Encompass
Catalog No. 50-971-986 Supplier Ace Glass, Inc. Supplier No. 570208