Learn More
Chemglass Life Sciences Cap, Bi-Metal, Magnetic, 20mm, Blue, White PTFE/Translucent Blue Silicone Ultra Low Bleed Septa
SDP
Catalog No. 501532540
Encompass_Preferred
Cap, Bi-Metal, Magnetic, 20mm, Blue, White PTFE/Translucent Blue Silicone Ultra Low Bleed Septa. The magnetic seals are for use with any instrument that transports the vial with the use of a magnet.
Catalog No. 50-153-2540 Supplier Chemglass Life Sciences Supplier No. CV32210020
Cap, Bi-Metal, Magnetic, 20mm, Blue, White PTFE/Translucent Blue Silicone Ultra Low Bleed Septa. The magnetic seals are for use with any instrument that transports the vial with the use of a magnet.