Learn More
Chemglass Life Sciences Open Top Cap, 53B, Polypropylene, Sterile - Individually bagged, 20 per case (fits 53B-style feed bottles)
SDP
Catalog No. NC2399408
Encompass
Sterile open-top polypropylene cap designed to close 53B feed bottles and compatible feed-bottle assemblies. Each cap is individually bagged and gamma sterilized, and is supplied 20 caps per case for laboratory and bioprocessing use.
  • Class VI polypropylene construction.
  • Individually bagged and gamma sterilized.
  • Supplied 20 per case for bulk laboratory use.
  • Open-top design for easy access and transfer applications.
Catalog No. NC2399408 Supplier Chemglass Life Sciences Supplier No. CLS11681053
Sterile open-top polypropylene cap designed to close 53B feed bottles and compatible feed-bottle assemblies. Each cap is individually bagged and gamma sterilized, and is supplied 20 caps per case for laboratory and bioprocessing use.
  • Class VI polypropylene construction.
  • Individually bagged and gamma sterilized.
  • Supplied 20 per case for bulk laboratory use.
  • Open-top design for easy access and transfer applications.