Learn More
Chemglass Life Sciences Cap, 70mm, Blue, Polypropylene, Non-Sterile
SDP
Catalog No. 50143773
Encompass_Preferred
Blue polypropylene cap is fully autoclavable at 121 degreesC.For a tight seal, cap must be used withsilicone liner (listed below).
Catalog No. 50-143-773 Supplier Chemglass Life Sciences Supplier No. CLS148770
Blue polypropylene cap is fully autoclavable at 121 degreesC.For a tight seal, cap must be used withsilicone liner (listed below).