Learn More
Quality Biological Inc 70mm Erlenmeyer/Fernbach Solid Cap, Cap Only, Sterile, 1/Bag, 24/CS
SDP
Catalog No. 50147476
Encompass_Preferred
70mm Erlenmeyer/Fernbach Solid Cap, CAP ONLY, Sterile
Packaged 1/Bag, 24/CS
• Replacement Cap for Erlenmeyer and Fernbach Flasks
Catalog No. 50-147-476 Supplier Quality Biological Inc Supplier No. P886229894
70mm Erlenmeyer/Fernbach Solid Cap, CAP ONLY, Sterile
Packaged 1/Bag, 24/CS
• Replacement Cap for Erlenmeyer and Fernbach Flasks