Learn More
Quality Biological Inc 70mm Erlenmeyer/Fernbach Vent Cap, Cap Only, Sterile, 1/Bag, 24/CS
SDP
Catalog No. 50147477
Encompass_Preferred
70mm Erlenmeyer/Fernbach Vent Cap, CAP ONLY, Sterile
Packaged 1/Bag, 24/CS
• Replacement Cap for Erlenmeyer and Fernbach Flasks
• Over-sized vent area in cap for improved gas exchange
Catalog No. 50-147-477 Supplier Quality Biological Inc Supplier No. P886229895
70mm Erlenmeyer/Fernbach Vent Cap, CAP ONLY, Sterile
Packaged 1/Bag, 24/CS
• Replacement Cap for Erlenmeyer and Fernbach Flasks
• Over-sized vent area in cap for improved gas exchange