Learn More
US Plastic Corporation 89/400 White Polypropylene Ribbed Cap with F217 Liner
SDP
Catalog No. NC1347048
Encompass
These caps are made of polypropylene and come with an F217 foamed polyethylene liner. They are known for good impact strength and flexibility, and they are cost-effective. Great for shampoo, lotion, and other hair care products.
This item is not returnable. View return policy
Catalog No. NC1347048 Supplier US Plastic Corporation Supplier No. 66506
These caps are made of polypropylene and come with an F217 foamed polyethylene liner. They are known for good impact strength and flexibility, and they are cost-effective. Great for shampoo, lotion, and other hair care products.