Learn More
Chemglass Life Sciences Injection molded cap assembly, GL45
SDP
Catalog No. 501531028
Encompass_Preferred
Injection molded cap assembly, GL45. Cap assembly fits GL45 threaded bottles and has a PTFE bulkhead with three 1/4in- 28 threaded ports to accomodate fittings for 1/8in O.D. tubing. Image shown is the fully assembled solvent reservoir.
Catalog No. 50-153-1028 Supplier Chemglass Life Sciences Supplier No. CG116616
Injection molded cap assembly, GL45. Cap assembly fits GL45 threaded bottles and has a PTFE bulkhead with three 1/4in- 28 threaded ports to accomodate fittings for 1/8in O.D. tubing. Image shown is the fully assembled solvent reservoir.