Learn More
Chemglass Life Sciences Cap Assembly, Complete, 1/8", GL-45, Red
SDP
Catalog No. 50871948
Encompass_Preferred
GL-45 Cap Assembly, Red, 1/8". An inexpensive bottle cap for either GL-45 or GPI-38 threads that permits standard solvent or media bottles to be used as an HPLC reservoir. Caps have two holes to accommodate 1/8" tubing and one hole that accepts a standard luer inner joint. Caps are made from polypropylene and the inserts are made from Tefzel. Complete cap assemblies consists of the cap, tefzel insert, (2) plugs for the 1/8" tubing openings and a plug for the opening.
Catalog No. 50-871-948 Supplier Chemglass Life Sciences Supplier No. CG115812
GL-45 Cap Assembly, Red, 1/8". An inexpensive bottle cap for either GL-45 or GPI-38 threads that permits standard solvent or media bottles to be used as an HPLC reservoir. Caps have two holes to accommodate 1/8" tubing and one hole that accepts a standard luer inner joint. Caps are made from polypropylene and the inserts are made from Tefzel. Complete cap assemblies consists of the cap, tefzel insert, (2) plugs for the 1/8" tubing openings and a plug for the opening.