Learn More
Waters Corp Waters Corporation Cap, Black, Phenolic, Screw Neck
Catalog No. 50785890 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Corporation offers a Black Phenolic Screw Neck Cap, designed for reliable laboratory use. This cap is compatible with most standard glass and plastic vials and provides a secure seal to prevent sample leakage. Its phenolic construction ensures heat resistance, insulation properties, and extreme resistance to chemical corrosion or low moisture adsorption.
  • Heat-resistant and insulation properties
  • Extremely resistant to chemical corrosion or low moisture adsorption
  • Compatible with most standard vials, including glass and plastic vials
  • Autoclavable for sterilization
  • Can be stored within a temperature range of -40°C to 100°C
  • Features a reliable sealing mechanism to prevent sample leakage
Catalog No. 50-785-890 Supplier Waters Corp Supplier No. WAT072711
Waters Corporation offers a Black Phenolic Screw Neck Cap, designed for reliable laboratory use. This cap is compatible with most standard glass and plastic vials and provides a secure seal to prevent sample leakage. Its phenolic construction ensures heat resistance, insulation properties, and extreme resistance to chemical corrosion or low moisture adsorption.
  • Heat-resistant and insulation properties
  • Extremely resistant to chemical corrosion or low moisture adsorption
  • Compatible with most standard vials, including glass and plastic vials
  • Autoclavable for sterilization
  • Can be stored within a temperature range of -40°C to 100°C
  • Features a reliable sealing mechanism to prevent sample leakage