Learn More
Waters Corp Waters Cap, Black, Phenolic, Screw Neck, 12 x 32 mm
Catalog No. 50785841 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This black phenolic screw neck cap provides a secure seal to prevent sample contamination and evaporation, making it suitable for both short-term and long-term sample storage. It is compatible with a wide range of laboratory solvents, including organic solvents, acids, and bases. The cap is autoclavable and can withstand temperatures up to 120°C, offering heat-resistant and insulation properties vital for samples at risk of chemical corrosion or low-moisture adsorption.
  • Screw neck closure type for a secure seal.
  • Black cap for easy identification.
  • Designed with 12 x 32 mm dimensions.
  • Includes both cap and septum.
  • Ideal for sample preparation applications.
  • Supplied in packages of 100.
  • Compatible with most automated sample handling systems.
Catalog No. 50-785-841 Supplier Waters Corp Supplier No. WAT058875
This black phenolic screw neck cap provides a secure seal to prevent sample contamination and evaporation, making it suitable for both short-term and long-term sample storage. It is compatible with a wide range of laboratory solvents, including organic solvents, acids, and bases. The cap is autoclavable and can withstand temperatures up to 120°C, offering heat-resistant and insulation properties vital for samples at risk of chemical corrosion or low-moisture adsorption.
  • Screw neck closure type for a secure seal.
  • Black cap for easy identification.
  • Designed with 12 x 32 mm dimensions.
  • Includes both cap and septum.
  • Ideal for sample preparation applications.
  • Supplied in packages of 100.
  • Compatible with most automated sample handling systems.