Learn More
Trajan Scientific 9 mm Blue Screw Cap w/ White/Silicone, Red/PTFE 1.0 mm thick septa, Pre-slit, Pack 100; LEAP PAL PARTS; mfg# LAP.09150869
Catalog No. NC1728142
Encompass_Preferred
9 mm Blue Screw Cap w/ White/Silicone, Red/PTFE 1.0 mm thick septa, Pre-slit, Pack 100; LEAP PAL PARTS; mfg# LAP.09150869
Catalog No. NC1728142 Supplier Trajan Scientific Supplier No. LAP.09150869
9 mm Blue Screw Cap w/ White/Silicone, Red/PTFE 1.0 mm thick septa, Pre-slit, Pack 100; LEAP PAL PARTS; mfg# LAP.09150869