Learn More
Chemglass Life Sciences Blue Polypropylene Cap, 20 mm, Pack of 100 (for Plain-End Culture Tubes)
SDP
Supplier:  Chemglass Life Sciences CLS150420B
Encompass_Preferred
Blue polypropylene cap designed for plain-end culture tubes. Molded from durable polypropylene and sold in packs of 100, the cap is autoclavable and features internal ridges to provide a secure grip and allow gas exchange, reducing evaporation during incubation and shaking applications.
  • Durable polypropylene construction, autoclavable to 121°C.
  • Fits plain-end culture tubes with a maximum diameter of 20 mm.
  • Internal ridges provide a secure grip and permit gas exchange.
  • Helps reduce sample evaporation during incubation and shaking.
  • Sold in packs of 100 for laboratory convenience.
Catalog No. NC2820465