Learn More
Agilent Technologies CAP, BLUE, SCREW, BONDED PRESL
Catalog No. NC2271161
Encompass
Cap, blue, screw, bonded preslit PTFE/silicone septa, 2 mL, 1000PK
This item is not returnable. View return policy
Catalog No. NC2271161 Supplier Agilent Technologies Supplier No. 51909067
Cap, blue, screw, bonded preslit PTFE/silicone septa, 2 mL, 1000PK