Learn More
Chemglass Life Sciences Cap, Bonded, Open Hole, Red, PTFE/Red Rubber, 9mm Thread
SDP
Catalog No. 501311403
Encompass_Preferred
Cap, Bonded, Open Hole, Red, PTFE/Red Rubber, 9mm Thread. Bonded closures feature a red rubber liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.
Catalog No. 50-131-1403 Supplier Chemglass Life Sciences Supplier No. CV3847R009
Cap, Bonded, Open Hole, Red, PTFE/Red Rubber, 9mm Thread. Bonded closures feature a red rubber liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.