Learn More
Beckman Coulter Cap Bottle, 63mm
Catalog No. NC1376089
Encompass_Preferred
The Fisherbrand 63mm Cap Bottle is a 63mm diameter plastic bottle with a screw cap closure. It has a capacity of 125mL and is made from low density polyethylene. The bottles are sold in cases of 500 and have graduations up to 100mL. The screw caps have a liner and an orifice diameter of 7.5mm. The bottles can withstand temperatures from -20°C to 80°C and the caps from -20°C to 60°C. The bottles conform to USP Type I requirements and meet FDA specifications for food contact.
Catalog No. NC1376089 Supplier Beckman Coulter Supplier No. 344693
The Fisherbrand 63mm Cap Bottle is a 63mm diameter plastic bottle with a screw cap closure. It has a capacity of 125mL and is made from low density polyethylene. The bottles are sold in cases of 500 and have graduations up to 100mL. The screw caps have a liner and an orifice diameter of 7.5mm. The bottles can withstand temperatures from -20°C to 80°C and the caps from -20°C to 60°C. The bottles conform to USP Type I requirements and meet FDA specifications for food contact.