Learn More
Cygnus Technologies CAP CELL LINE HCP ELISA KIT
Supplier:  Cygnus Technologies F820
Encompass
CAP Cell Line HCP ELISA Kit (F820)The CAP cell line, being of human origin, provides recombinant proteins with human-type glycosylation and is commonly employed in the production of recombinant proteins, gene therapy products, and vaccines. CAP cell line is a CEVEC Pharmaceuticals GmbH core proprietary technology. Our kit has been qualified for purification process and in-process lot release testing. This kit has an LOD of ~0.9 ng/ml.
Catalog No. NC1813896