Learn More
Ace Glass, Inc. CAP CENTER HOLE GL18 PBT CAP CENTER HOLE GL18 PBT
SDP
Catalog No. 50975135
Encompass
GL18 Duran, polybutylene terephthalate (PBT) open-top caps with center hole. PBT temperature range is -45 - 180¿C. Can be used with Duran hose connectors, Duran sealing ring for GL thread hose connectors, and silicone or silicone/PTFE septa for Duran caps.
Catalog No. 50-975-135 Supplier Ace Glass, Inc. Supplier No. 762108
GL18 Duran, polybutylene terephthalate (PBT) open-top caps with center hole. PBT temperature range is -45 - 180¿C. Can be used with Duran hose connectors, Duran sealing ring for GL thread hose connectors, and silicone or silicone/PTFE septa for Duran caps.