Learn More
Ace Glass, Inc. CAP CENTER HOLE GL32 PBT CAP CENTER HOLE GL32 PBT
SDP
Catalog No. 50975137
Encompass
GL32 Duran, polybutylene terephthalate (PBT) open-top caps with center hole. PBT temperature range is -45 - 180¿C. Can be used with Duran hose connectors, Duran sealing ring for GL thread hose connectors, and silicone or silicone/PTFE septa for Duran caps.
Catalog No. 50-975-137 Supplier Ace Glass, Inc. Supplier No. 762122
GL32 Duran, polybutylene terephthalate (PBT) open-top caps with center hole. PBT temperature range is -45 - 180¿C. Can be used with Duran hose connectors, Duran sealing ring for GL thread hose connectors, and silicone or silicone/PTFE septa for Duran caps.