Learn More
Chemglass Life Sciences Cap, Chem Snap, 11mm, Blue, Pre-Cut Septa, PTFE/Silicone
SDP
Catalog No. 501532581
Encompass_Preferred
Cap, Chem Snap, 11mm, Blue, Pre-Cut Septa, PTFE/Silicone. Chem Snap Caps include pre-cut septa where only the carrier is cut through in a Y-form, leaving the PTFE lamination intact. The unhurt PTFE lamination gives 100% protection to the sample while leaving a 0.05mm thick PTFE portion remaining for needle penetration. The Y-form in the pre-cut silicone allows for a better flow of air during sample pick up in order to avoid a vacuum in the vial.
Catalog No. 50-153-2581 Supplier Chemglass Life Sciences Supplier No. CV3844B011
Cap, Chem Snap, 11mm, Blue, Pre-Cut Septa, PTFE/Silicone. Chem Snap Caps include pre-cut septa where only the carrier is cut through in a Y-form, leaving the PTFE lamination intact. The unhurt PTFE lamination gives 100% protection to the sample while leaving a 0.05mm thick PTFE portion remaining for needle penetration. The Y-form in the pre-cut silicone allows for a better flow of air during sample pick up in order to avoid a vacuum in the vial.