Learn More
Cellscript Inc Cap-Clip™ Acid Pyrophosphatase | 100 units
Catalog No. NC1723919
Encompass
For the removal of cap structures (decapping) from various RNAs (i.e. mRNAs) resulting in 5'-monophosphate RNAs. Can be substituted unit for unit, into any preexisting protocol for Tobacco Acid Pyrophosphatase (TAP) utilizing the same buffer.
Catalog No. NC1723919 Supplier Cellscript Inc Supplier No. CCC15011H
For the removal of cap structures (decapping) from various RNAs (i.e. mRNAs) resulting in 5'-monophosphate RNAs. Can be substituted unit for unit, into any preexisting protocol for Tobacco Acid Pyrophosphatase (TAP) utilizing the same buffer.