Learn More
YMC America, Inc Cap Coupler 1/16in OD 0.1mm ID
SDP
Catalog No. 5000876542
Encompass
Capillary column coupler 1/16 OD 0 1 mm ID 10-32 thread Includes a stainless steel tube 2 male nuts and 2 ferrules
Catalog No. 50-008-76542 Supplier YMC America, Inc Supplier No. XRCP1601E1PK
Capillary column coupler 1/16 OD 0 1 mm ID 10-32 thread Includes a stainless steel tube 2 male nuts and 2 ferrules