Learn More
YMC America, Inc Cap Coupler 1/32in OD 0.13mmID
SDP
Supplier:  YMC America, Inc XRCP3201E1PK
Encompass
Capillary column coupler 1/32 OD 0 13 mm ID 6-40 thread Includes a stainless steel tube 2 male nuts and 2 ferrules
Catalog No. 50-008-76383