Learn More
Microsolv Technology Corporation Cap Crimp 8Mm Nat Rub/Tef
Catalog No. 5001452248
Encompass_Preferred
Caps, Crimp Top, with 1.0mm thick, Red-Orange Natural Rubber / Transparent TEF Septa, fitted in aluminum, Silver Caps with a 4mm center hole. For use with 8mm Crimp Top autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52248 Supplier Microsolv Technology Corporation Supplier No. 9508C40TS
Caps, Crimp Top, with 1.0mm thick, Red-Orange Natural Rubber / Transparent TEF Septa, fitted in aluminum, Silver Caps with a 4mm center hole. For use with 8mm Crimp Top autosampler vials. MicroSolv Brand. 100/PK.