Learn More
Microsolv Technology Corporation Cap Crimp 8Mm Sil/Ptfe Ultra
Catalog No. 5001452246
Encompass_Preferred
Caps, Crimp Top, with 1.3mm thick, White Silicone Rubber / Red PTFE, ultra-clean Septa, fitted in aluminum, Silver Caps with a 4mm center hole. For use with 8mm Crimp Top autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52246 Supplier Microsolv Technology Corporation Supplier No. 9508C10TS
Caps, Crimp Top, with 1.3mm thick, White Silicone Rubber / Red PTFE, ultra-clean Septa, fitted in aluminum, Silver Caps with a 4mm center hole. For use with 8mm Crimp Top autosampler vials. MicroSolv Brand. 100/PK.