Learn More
Microsolv Technology Corporation Cap Crimp Gold Red Rubbercap
Catalog No. 5001451971
Encompass_Preferred
Caps, Crimp Top, with Red Rubber / PTFE Septa, fitted in aluminum, Gold Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. BASIK Brand. 100/PK.
Catalog No. 50-014-51971 Supplier Microsolv Technology Corporation Supplier No. 9502C40TD
Caps, Crimp Top, with Red Rubber / PTFE Septa, fitted in aluminum, Gold Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. BASIK Brand. 100/PK.