Learn More
Trajan Scientific L-MARK Brand Cap Crimp 20 mm Headspace Tan PTFE/White Silicone, 100 Pack
Catalog No. 501090133
Encompass_Preferred
L-MARK Brand Cap Crimp Aluminum 20mm Tan PTFE/White Silicone 3.2mm thick Standard Headspace 100 Pack. Made in the USA.
Catalog No. 50-109-0133 Supplier Trajan Scientific Supplier No. LPP.020039982
L-MARK Brand Cap Crimp Aluminum 20mm Tan PTFE/White Silicone 3.2mm thick Standard Headspace 100 Pack. Made in the USA.