Learn More
Trajan Scientific La-Pha-Pack , Cap, Crimp, Headspace, 20mm, Magnetic, Butyl 3.0mm septa, 100 Pack. Made in Germany.
Catalog No. 501037352
Encompass_Preferred
La-Pha-Pack , Cap, Crimp, Headspace, 20mm, Magnetic, Butyl 3.0mm septa, 100 Pack. Made in Germany.
Catalog No. 50-103-7352 Supplier Trajan Scientific Supplier No. LAP.20031157
La-Pha-Pack , Cap, Crimp, Headspace, 20mm, Magnetic, Butyl 3.0mm septa, 100 Pack. Made in Germany.