Learn More
Microsolv Technology Corporation Cap Crimp Top Magnetic Goldca
Catalog No. 5001451945
Encompass_Preferred
Caps, Crimp Top, with 1.0mm thick, Red PTFE / White Silicone Rubber / Red PTFE Septa, fitted in steel, magnetic Gold Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51945 Supplier Microsolv Technology Corporation Supplier No. 9502C20MD
Caps, Crimp Top, with 1.0mm thick, Red PTFE / White Silicone Rubber / Red PTFE Septa, fitted in steel, magnetic Gold Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.