Learn More
Trajan Scientific L-MARK Brand Cap Crimp 20 mm Headspace Tan PTFE/White Silicone Press Release, Case of 1000
Catalog No. 501251009
Encompass_Preferred
L-MARK Brand, Cap, Crimp, Aluminum, 20mm, Tan PTFE/White Silicone, 3.2mm thick, Pressure Release, Headspace, Case of 1000. Made in the USA.
Catalog No. 50-125-1009 Supplier Trajan Scientific Supplier No. LPP.02003960710
L-MARK Brand, Cap, Crimp, Aluminum, 20mm, Tan PTFE/White Silicone, 3.2mm thick, Pressure Release, Headspace, Case of 1000. Made in the USA.