Learn More
Trajan Scientific L-MARK Brand Cap Crimp 20 mm Headspace Tan PTFE/White Silicone, Case of 1000
Catalog No. 501251012
Encompass_Preferred
L-MARK Brand Cap Crimp Aluminum 20mm Tan PTFE/White Silicone 3.2mm thick Standard Headspace 1000/case. Made in the USA.
Catalog No. 50-125-1012 Supplier Trajan Scientific Supplier No. LPP.02003998210
L-MARK Brand Cap Crimp Aluminum 20mm Tan PTFE/White Silicone 3.2mm thick Standard Headspace 1000/case. Made in the USA.