Learn More
Hach Company Storage Caps for Rugged PHC and MTC Electrode, 5 pcs
Supplier:  Hach Company 5857305
Encompass
Pack of 5 caps for short-term and long-term storage of the Rugged PHC101 & MTC101 probes. Using cap for storage prevents reference junction from dry out to ensure best performances.
Catalog No. 50-199-7197