Learn More
IDEX Health & Science Upchurch CAP F/1/16IN MALE NUT TEFZEL
Catalog No. NC1919178
Encompass_Preferred
Idex Threaded Plugs and Caps, Yellow, Tefzel (ETFE), Port Type: 5/16-24
Catalog No. NC1919178 Supplier IDEX Health & Science Upchurch Supplier No. P754
Idex Threaded Plugs and Caps, Yellow, Tefzel (ETFE), Port Type: 5/16-24