Learn More
Brookfield Engineer Labs CAP FLUID
Catalog No. NC9967784
Encompass
3542 CPS Mineral Oil Standard for High Temp High Torque CAP with Cone 6 Spindle
Catalog No. NC9967784 Supplier Brookfield Engineer Labs Supplier No. CAP6H
3542 CPS Mineral Oil Standard for High Temp High Torque CAP with Cone 6 Spindle