Learn More
Microsolv Technology Corporation Cap For 40Ml Epa Vials Solid
Catalog No. 5001452444
Encompass_Preferred
Caps, Screw Top, Solid, with Red Butyl Rubber / Grey PTFE Liner, fitted in knurled polypropylene, White Caps. For use with 24-400mm threaded EPA Style vials. ND24 threads. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52444 Supplier Microsolv Technology Corporation Supplier No. 97040SP24
Caps, Screw Top, Solid, with Red Butyl Rubber / Grey PTFE Liner, fitted in knurled polypropylene, White Caps. For use with 24-400mm threaded EPA Style vials. ND24 threads. MicroSolv Brand. 100/PK.