Learn More
Chemglass Life Sciences Cap, GL-25, Blue, Solid, Polypropylene
SDP
Catalog No. 501215127
Encompass_Preferred
Solid polypropylene caps with a molded in lip seal.All color caps can be autoclaved in steam to 140 degreesC.
Catalog No. 50-121-5127 Supplier Chemglass Life Sciences Supplier No. CLS148010
Solid polypropylene caps with a molded in lip seal.All color caps can be autoclaved in steam to 140 degreesC.