Learn More
Chemglass Life Sciences Cap, GL-32, Blue, Solid, Polypropylene
SDP
Supplier:  Chemglass Life Sciences CLS148011
Encompass_Preferred
Solid polypropylene caps with a molded in lip seal.All color caps can be autoclaved in steam to 140 degreesC.
Catalog No. 50-000-83255