Learn More
Chemglass Life Sciences Cap, GL-32, Orange, Solid, Polypropylene
SDP
Catalog No. 501215129
Encompass_Preferred
Solid polypropylene caps with a molded in lip seal.All color caps can be autoclaved in steam to 140 degreesC.
Catalog No. 50-121-5129 Supplier Chemglass Life Sciences Supplier No. CLS148012
Solid polypropylene caps with a molded in lip seal.All color caps can be autoclaved in steam to 140 degreesC.